


Cyanogen bromide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:506-68-3
:
Formula:CBrN
MDL Number:MFCD00011597
:105.92 g/mol
Merck Index:12,02763
Boiling Pt:61.4 °C (1013 hPa)
Melting Pt:52 °C
Density:2.06 g/cm³ (20 °C)
Storage Temperature:Refrigerator
UN:1889
ADR:6.1,I
:M-Clarity™ quality level = MQ200
EINECS:208-051-2
:GHS06
:GHS05
:GHS09
Frequently Bought Together