Order Entry
Czech Republic
ContactUsLinkComponent
Cyanogen bromide, Sigma-Aldrich®
Toxic
Corrosive
Environment
Cyanogen bromide, Sigma-Aldrich®
Catalog # SIAL481432-25G
Supplier:  Merck
CAS Number:  
Toxic
Corrosive
Environment
Cyanogen bromide, Sigma-Aldrich®
Catalog # SIAL481432-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :105.92 g/mol
Merck Index:12,02763
Boiling Pt:61.4 °C (1013 hPa)
Melting Pt:52 °C
Density:2.06 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:208-051-2
UN:1889
ADR:6.1,I
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS05
 :GHS09
CAS Number:506-68-3
Formula:CBrN
MDL Number:MFCD00011597