Order Entry
Czech Republic
ContactUsLinkComponent
Iodine trichloride (Wijs'reagent) ≥97%, Sigma-Aldrich®
Corrosive
Irritant
Iodine trichloride (Wijs'reagent) ≥97%, Sigma-Aldrich®
Catalog # SIAL359122-50G
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Iodine trichloride (Wijs'reagent) ≥97%, Sigma-Aldrich®
Catalog # SIAL359122-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :233.26 g/mol
Merck Index:13,05042
Melting Pt:63 °C
Density:1.23 g/cm³ (20 °C)
Flash Pt:40 °C
Storage Temperature:Freezer
EINECS:212-739-8
UN:2920
ADR:8,II
 :M-Clarity™ quality level = MQ100
 :GHS05
 :GHS07
CAS Number:865-44-1
Formula:ICl₃
MDL Number:MFCD00036291