Iodine monochloride, Sigma-Aldrich®
: Merck
:7790-99-0
:
:ClI
:MFCD00011354
:162.36 g/mol
:13,05039
:97.3 °C (1013 hPa)
:26 °C
:3.1 g/cm³ (29 °C)
:1792
:8,II
:M-Clarity™ quality level = MQ100
:232-236-7
:GHS05
:GHS07
:Ambient
Frequently Bought Together