

Iodine monochloride for synthesis, Sigma-Aldrich®
: Merck
:7790-99-0
:
:ClI
:MFCD00011354
:162.36 g/mol
:13,05039
:97.3 °C (1013 hPa)
:26 °C
:3.1 g/cm³ (29 °C)
:3498
:8,II
:M-Clarity™ quality level = MQ200
:232-236-7
:GHS05
:GHS07
:Ambient
- Solubility:(20 °C) (decomposition)
- Melting Point:14 - 27 °C
- Molar Mass:162.35 g/mol
- Boiling Point:97 °C (1013 hPa) (decomposition)
- Density:3.1 g/cm3 (29 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Frequently Bought Together