

Iodine monochloride
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7790-99-0
:
Formula:ClI
MDL Number:MFCD00011354
:162.36 g/mol
Merck Index:14,5017
Boiling Pt:96…98 °C
Melting Pt:25…27 °C
Density:3.1822
Storage Temperature:Ambient
EINECS:232-236-7
UN:1792
ADR:8,II
:GHS05
:GHS07
Related Information
Frequently Bought Together