Order Entry
Czech Republic
ContactUsLinkComponent
Iodine monobromide solution 0.1 mol/l (0.2 N) in acetic acid according to Hanus for the determination of iodine adsorption number
Corrosive
Irritant
Iodine monobromide solution 0.1 mol/l (0.2 N) in acetic acid according to Hanus for the determination of iodine adsorption number
Catalog # VWRS5527.0100
Supplier:  VWR Collection
CAS Number:  
Corrosive
Irritant
Iodine monobromide solution 0.1 mol/l (0.2 N) in acetic acid according to Hanus for the determination of iodine adsorption number
Catalog # VWRS5527.0100
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :206.81 g/mol
Merck Index:13,05038
Boiling Pt:116 °C (1013 hPa)
Melting Pt:37…40 °C
Density:4.41 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-159-9
UN:3260
ADR:8,II
 :GHS05
 :GHS07
CAS Number:7789-33-5
Formula:IBr
MDL Number:MFCD00011353