2,2'-Dipyridyl disulphide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2127-03-9
:
Formula:C₁₀H₈N₂S₂
MDL Number:MFCD00006287
:220.32 g/mol
Boiling Pt:348 °C (1013 hPa)
Melting Pt:57…61 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
EINECS:218-343-1
Frequently Bought Together