Order Entry
Czech Republic
ContactUsLinkComponent
Dimethyl disulphide for synthesis, Sigma-Aldrich®
Dimethyl disulphide for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Dimethyl disulphide for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    2.5 g/l (20 °C)
  • Melting Point:
    -85 °C
  • Molar Mass:
    94.20 g/mol
  • Boiling Point:
    110 °C (1013 hPa)
  • Vapor Pressure:
    28 hPa (20 °C)
  • Flash Point:
    16 °C
  • Refractive Index:
    1.525 (20 °C, 589 nm)
  • Explosion Limit:
    1.1 - 16.1 %(V)
  • Density:
    1.06 g/cm3 (20 °C)
  • Ignition Point:
    370 °C
  • Synonyms:
    Methyl disulfide
  • Hill Formula:
    C2H6S2
  • Chemical Formula:
    CH3SSCH3

Specification Test Results

Related Information

Frequently Bought Together