Diethyl sulphide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:352-93-2
:
Formula:C₄H₁₀S
MDL Number:MFCD00009270
:90.19 g/mol
Merck Index:13,03885
Boiling Pt:92 °C (1013 hPa)
Melting Pt:–104 °C
Density:0.837 g/cm³ (20 °C)
UN:2375
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:206-526-9
Frequently Bought Together