Dibutyl sulphide for synthesis, Sigma-Aldrich®
: Merck
:544-40-1
:
:C₈H₁₈S
:MFCD00009468
:146.3 g/mol
:12,01626
:189 °C (1013 hPa)
:–80 °C
:0.83 g/cm³ (20 °C)
:Ambient
:208-870-5
:3334
:9,
:M-Clarity™ quality level = MQ200
:62 °C
- Solubility:(20 °C) insoluble
- Melting Point:-80 °C
- Molar Mass:146.29 g/mol
- Boiling Point:188.7 °C (1013 hPa)
- Vapor Pressure:26 hPa (80 °C)
- Flash Point:62 °C
- Refractive Index:1.453 (20 °C, 589 nm)
- Density:0.84 g/cm3 (20 °C)
- Synonyms:Butyl sulfide
- Hill Formula:C8H18S
- Chemical Formula:CH3(CH2)3S(CH2)3CH3
Frequently Bought Together