
Dibutyl disulphide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:629-45-8
:
Formula:C₈H₁₈S₂
MDL Number:MFCD00009467
:178.36 g/mol
Boiling Pt:229…233 °C (760 mmHg)
Melting Pt:-94 °C
Density:0.938 g/cm³
Flash Pt:93 °C
EINECS:211-091-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4915 to 1.4935 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together