
Dibutyl disulphide
Catalog # ABCRAB141083.0025
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
Specifications
About this item
:
:178.36 g/mol
Boiling Pt:229 °C (1013 hPa)
Melting Pt:–94 °C
Density:0,9371 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-091-3
:GHS07
CAS Number:629-45-8
Formula:C₈H₁₈S₂
MDL Number:MFCD00009467