
Thiodiglycol ≥99%, TECHNICAL
Catalog # 28620.182
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Assay:Min. 99 %
Specification Test Results
About this item
:
:122.19 g/mol
Merck Index:12,09466
Boiling Pt:282 °C (1013 hPa)
Melting Pt:–10 °C
Density:1.18 g/cm³ (20 °C)
Flash Pt:113 °C (closed cup)
EINECS:203-874-3
UN:3334
ADR:,
:GHS07
CAS Number:111-48-8
Formula:S(CH₂CH₂OH)₂