Order Entry
Czech Republic
ContactUsLinkComponent
Dimethyl sulphoxide, anhydrous
Dimethyl sulphoxide, anhydrous
Catalog # BKC-17
Supplier:  G-Biosciences
CAS Number:  
undefined
Dimethyl sulphoxide, anhydrous
Catalog # BKC-17
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
 :
 :78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:200-664-3
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089