


Thionyl chloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7719-09-7
:
Formula:Cl₂OS
MDL Number:MFCD00011449
:118.97 g/mol
Merck Index:12,09485
Boiling Pt:76 °C (1013 hPa)
Melting Pt:–104.5 °C
Density:1.64 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1836
ADR:8,I
:M-Clarity™ quality level = MQ200
EINECS:231-748-8
:GHS06
:GHS05
:GHS07
Specification Test Results
- Solubility:(20 °C) (rigorous decomposition)
- Melting Point:-104.5 °C
- Molar Mass:118.97 g/mol
- Boiling Point:76 °C (1013 hPa)
- Vapor Pressure:121 hPa (20 °C)
- Refractive Index:1.517 (20 °C, 589 nm)
- Density:1.64 g/cm3 (20 °C)
- pH:(H2O) not applicable
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Frequently Bought Together