Order Entry
Czech Republic
ContactUsLinkComponent
Disulphur dichloride for synthesis, Sigma-Aldrich®
Toxic
Corrosive
Environment
Disulphur dichloride for synthesis, Sigma-Aldrich®
Catalog # 8.14179.1000
Supplier:  Merck
CAS Number:  
Toxic
Corrosive
Environment
Disulphur dichloride for synthesis, Sigma-Aldrich®
Catalog # 8.14179.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    (20 °C) (rigorous decomposition)
  • Melting Point:
    -76 °C
  • Molar Mass:
    135.04 g/mol
  • Boiling Point:
    138 °C
  • Vapor Pressure:
    14.7 hPa (20 °C)
  • Flash Point:
    4.2 - 32.5 %(V)
  • Refractive Index:
    130 °C
  • Explosion Limit:
    1.67 (20 °C, 589 nm)
  • Density:
    1.68 g/cm3 (20 °C)
  • Ignition Point:
    390 °C
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
 :135.04 g/mol
Merck Index:12,09143
Boiling Pt:138 °C (1013 hPa)
Melting Pt:–76 °C
Density:1.68 g/cm³ (20 °C)
Flash Pt:118 °C
Storage Temperature:Ambient
EINECS:233-036-2
UN:1828
ADR:8,I
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS05
 :GHS09
CAS Number:10025-67-9
Formula:Cl₂S₂