
Ammonium hexafluorosilicate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16919-19-0
:
Formula:H₈F₆N₂Si
MDL Number:MFCD00010887
Merck Index:14,527
Melting Pt: Dec.
Density:2.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:240-968-3
UN:2854
ADR:6.1,III
:178.15 g/mol
:GHS06
Related Information
Frequently Bought Together