Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Ammonium thiocyanate ≥99% (by argentometric titration), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
 
 
Ammonium thiocyanate ≥99% (by argentometric titration), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
 

 

  • Assay (argentometric):
    Min. 99.0 %
  • Water insoluble matter:
    Max. 0.005 %
  • Residue on ignition (SO₄):
    Max. 0.02 %
  • pH (5 %, 25 °C):
    4.5 - 6.0
  • Lead (Pb):
    Max. 0.0004 %
  • Iron (Fe):
    Max. 1 ppm
  • Copper (Cu):
    Max. 0.0005 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 0.004 %
  • Sulfate (SO₄):
    Max. 0.005 %
  • Sulfide (S):
    Max. 0.001 %
  • I reducing matters (as SO₂):
    Max. 0.005 %

 

 

Frequently Bought Together