Order Entry
Czech Republic
ContactUsLinkComponent
Ammonium dihydrogen phosphate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Ammonium dihydrogen phosphate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Ammonium dihydrogen phosphate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    368 g/l (20 °C)
  • Melting Point:
    190 °C
  • Molar Mass:
    115.02 g/mol
  • Bulk Density:
    800 - 1100 kg/m3
  • Vapor Pressure:
    0.066 hPa (125 °C)
  • Density:
    1.80 g/cm3 (20 °C)
  • pH:
    3.8 - 4.4 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together