
Ammonium iodide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:12027-06-4
:
Formula:NH₄I
MDL Number:MFCD00011424
Merck Index:13,00531
Melting Pt:551 °C
Density:2.51 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:144.94 g/mol
EINECS:234-717-7
:GHS07
Storage Temperature:Ambient
Frequently Bought Together