
Ammonium fluoride, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:12125-01-8
:
Formula:NH₄F
MDL Number:MFCD00011423
:37.04 g/mol
Merck Index:13,00524
Melting Pt:160 °C
Density:1.0088…1.0096 g/cm³ (25 °C)
EINECS:235-185-9
ADR:6.1,III
:M-Clarity™ quality level = MQ300
UN:2505
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Solubility:820 g/l (20 °C)
- Molar Mass:37.04 g/mol
- Bulk Density:250 - 350 kg/m3
- Density:1.01 g/cm3 (20 °C)
- pH:6 (50 g/l, H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_101164
SDS
ProductPage_101164 ProductPage_101164
Product specification
Frequently Bought Together