Order Entry
Czech Republic
ContactUsLinkComponent
Ammonium carbonate, extra pure
Irritant
Ammonium carbonate, extra pure
Certificates
undefined
Irritant
Ammonium carbonate, extra pure
Certificates

Specification Test Results

  • Appearance:
    White Powder or crystals and/or chunks
  • Non-volatile matter:
    ≤100 ppm
  • Titration with HCl:
    ≥30.5 % (NH3)
  • Chloride (Cl):
    ≤30 ppm
  • Sulfate (SO4):
    ≤30 ppm
  • Iron (Fe):
    ≤5 ppm
  • Heavy metals (as Pb):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together