Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Ammonium bromide, EMSURE® ACS for analysis, Supelco®
Catalog # 1.01125.1000
Supplier:  Merck
CAS Number:  
Irritant
Ammonium bromide, EMSURE® ACS for analysis, Supelco®
Catalog # 1.01125.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Solubility:
    742 g/l (20 °C)
  • Melting Point:
    542 °C (closed capillary tube)
  • Molar Mass:
    97.94 g/mol
  • Bulk Density:
    1100 kg/m3
  • Vapor Pressure:
    1.3 hPa (198 °C)
  • Density:
    2.43 g/cm3 (20 °C)
  • pH:
    4.8 (50 g/l, H2O, 25 °C)

Specification Test Results

About this item

 :
 :97.94 g/mol
Merck Index:13,00505
Melting Pt:542 °C
Density:2.42 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ300
EINECS:235-183-8
 :GHS07
Storage Temperature:Ambient
CAS Number:12124-97-9
Formula:H₄BrN
MDL Number:MFCD00011419