
Tetraphenylphosphonium chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2001-45-8
:
Formula:C₂₄H₂₀ClP
MDL Number:MFCD00011916
:374.85 g/mol
Melting Pt:275…280 °C
EINECS:217-890-3
:GHS07
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:275 to 280 °C
- Titration Argentometric:≥97.5 %
Specification Test Results
Frequently Bought Together