

Triphenylphosphine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:603-35-0
:
Formula:C₁₈H₁₅P
MDL Number:MFCD00003043
:262.29 g/mol
Merck Index:12,09873
Boiling Pt:377 °C (1013 hPa)
Melting Pt:79 °C
Density:1.194 g/cm³ (20 °C)
Flash Pt:180 °C
Storage Temperature:Ambient
EINECS:210-036-0
:GHS07
:GHS09
Frequently Bought Together