
Tris(4-methylphenyl)phosphine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1038-95-5
:
Formula:C₂₁H₂₁P
MDL Number:MFCD00008542
:304.37 g/mol
Boiling Pt:400 °C (1013 hPa)
Melting Pt:145…150 °C
Density:1,11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:213-863-5
:GHS07
Specification Test Results
- Appearance:White to light yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:144 to 148 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together