

Dichloro(phenyl)phosphine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:644-97-3
:
Formula:C₆H₅Cl₂P
MDL Number:MFCD00000528
:178.98 g/mol
Boiling Pt:222 °C (1013 hPa)
Melting Pt:–51 °C
Density:1.32 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-425-8
ADR:8,II
:M-Clarity™ quality level = MQ100
UN:2798
:GHS06
:GHS05
Frequently Bought Together