Zadání objednávky
Czech Republic
ContactUsLinkComponent
Nitronium tetrafluoroborate 97%
Corrosive
Nitronium tetrafluoroborate 97%
Certificates
undefined
Corrosive
Nitronium tetrafluoroborate 97%

Specification Test Results

  • Appearance:
    Colorless to white Adhering crystals
  • Assay:
    ≥96.0 %
  • Melting point:
    56 to 63 °C
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together