Order Entry
Czech Republic
ContactUsLinkComponent
Nitronium tetrafluoroborate
Nitronium tetrafluoroborate
Catalog # ABCRF05270
Supplier:  abcr
CAS Number:  
Nitronium tetrafluoroborate
Catalog # ABCRF05270
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :132.81 g/mol
Melting Pt:260 °C
Density:1.26 g/cm³ (20 °C)
EINECS:237-533-5
UN:3260
ADR:8,II
CAS Number:13826-86-3
Formula:BF₄NO₂
MDL Number:MFCD00011432