Order Entry
Czech Republic
ContactUsLinkComponent
Dicyclohexylphosphine ≥98%
Flammable
Toxic
Dicyclohexylphosphine ≥98%
Catalog # ABCRAB112645.0010
Supplier:  abcr
CAS Number:  
Flammable
Toxic
Dicyclohexylphosphine ≥98%
Catalog # ABCRAB112645.0010
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :198.29 g/mol
Boiling Pt:129 °C (8 mmHg)
Density:0.98
EINECS:212-590-9
UN:2845
ADR:4.2,I
 :GHS02
 :GHS06
CAS Number:829-84-5
Formula:C₁₂H₂₃P
MDL Number:MFCD00014296