Butyrylthiocholine iodide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1866-16-6
:
Formula:H₃C(CH₂)₂C(S)O(CH₂)₂N(CH₃)₃I
MDL Number:MFCD00011845
:317.23 g/mol
Melting Pt:172 °C
:M-Clarity™ quality level = MQ300
Storage Temperature:Freezer
EINECS:217-475-7
Frequently Bought Together