


Tetramethylammonium chloride for synthesis, Sigma-Aldrich®
8.22156.1000
: Merck
:
- Pack type:Glass bottle
- Pk:1 kg
- Seq:4024
- Solubility:(20 °C) soluble
- Melting Point:300 °C
- Molar Mass:109.60 g/mol
- Vapor Pressure:(20 °C)
- Flash Point:100 °C
- Density:1.17 g/cm3 (20 °C)
- pH:6 - 8 (100 g/l, H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
:
:109.60 g/mol
:> 300 °C
:1.169 g/cm³ (20 °C)
:2811
:6.1,II
:M-Clarity™ quality level = MQ200
:>100 °C
:200-880-8
:GHS06
:GHS08
:GHS09
:Ambient
:75-57-0
:(CH₃)₄NCl
:MFCD00011628