
Tetrahexylammonium bromide, Supelco®
Catalog # 87297-10G-F
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:10 g
- Sekvence:3236
Specifications
About this item
:
Melting Pt:104 °C
Density:1,0036 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ100
EINECS:224-363-1
:GHS07
CAS Number:4328-13-6
Formula:C₂₄H₅₂BrN
MDL Number:MFCD00011858