
Tetraheptylammonium bromide 99%
:4368-51-8
:
:C₂₈H₆₀BrN
:MFCD00011861
:490.7 g/mol
:89…91 °C
:224-459-3
:GHS07
:Ambient
- Appearance:White Flakes
- Infrared spectrum:Conforms
- Melting point:89 to 91 °C
- Titration Argentometric:≥98.5 %
Frequently Bought Together