Order Entry
Czech Republic
ContactUsLinkComponent
Cetrimonium bromide for synthesis, Sigma-Aldrich®
Cetrimonium bromide for synthesis, Sigma-Aldrich®
Catalog # 8.14119.0500
Supplier:  Merck
CAS Number:  
Cetrimonium bromide for synthesis, Sigma-Aldrich®
Catalog # 8.14119.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Solubility:
    0.192 g/l (20 °C)
  • Melting Point:
    250 - 256 °C
  • Molar Mass:
    364.45 g/mol
  • Bulk Density:
    390 kg/m3
  • pH:
    5 - 7 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    9
  • Hazard Class Description:
    Miscellaneous Hazardous Material

Specification Test Results

About this item

 :
 :364.45 g/mol
Merck Index:13,02034
Melting Pt:237…243 °C
UN:3077
ADR:9,III
 :M-Clarity™ quality level = MQ200
Flash Pt:244 °C (closed cup)
EINECS:200-311-3
 :GHS05
 :GHS08
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:57-09-0
Formula:CH₃(CH₂)₁₅N(Br)(CH₃)₃
MDL Number:MFCD00011772