Order Entry
Czech Republic
ContactUsLinkComponent
Cetrimonium bromide ≥99%, high purity
Corrosive
Irritant
Environment
Cetrimonium bromide ≥99%, high purity
Catalog # 0833-1KG
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Irritant
Environment
Cetrimonium bromide ≥99%, high purity
Catalog # 0833-1KG
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Purity:
    ≥99%

Specification Test Results

About this item

 :
 :364.45 g/mol
Merck Index:13,02034
Melting Pt:237…243 °C
Flash Pt:244 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-311-3
UN:3077
ADR:9,III
 :GHS05
 :GHS07
 :GHS09
CAS Number:57-09-0
Formula:CH₃(CH₂)₁₅N(Br)(CH₃)₃
MDL Number:MFCD00011772