Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
(Methyl)triphenylphosphonium bromide 98%
 
undefined
 
 
(Methyl)triphenylphosphonium bromide 98%

 

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    230 to 234 °C
  • Titration Argentometric:
    ≥97.5 %

 

 

Frequently Bought Together