
Diethyl methylphosphonate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:683-08-9
:
Formula:C₅H₁₃O₃P
MDL Number:MFCD00009813
:152.13 g/mol
Boiling Pt:194 °C
Density:1.040
Flash Pt:75 °C (167 °F)
EINECS:211-667-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together