Trimethyl phosphite, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:121-45-9
:
Formula:C₃H₉O₃P
MDL Number:MFCD00008350
:124.08 g/mol
Boiling Pt:111 °C (1013 hPa)
Melting Pt:–77.8 °C
Density:1.042 g/cm³ (20 °C)
Flash Pt:50 °C
UN:2329
ADR:3,III
:M-Clarity™ quality level = MQ200
EINECS:204-471-5
Frequently Bought Together