Trimethyl phosphite for synthesis, Sigma-Aldrich®
Catalog # 8.00553.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Melting Point:-78 °C
- Molar Mass:124.08 g/mol
- Boiling Point:112 °C (1013 hPa)
- Vapor Pressure:28 hPa (20 °C)
- Flash Point:28 °C
- Refractive Index:1.408 (20 °C, 589 nm)
- Explosion Limit:5.2 - 61.2 %(V)
- Density:1.05 g/cm3 (20 °C)
- Ignition Point:250 °C
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
About this item
:
:124.08 g/mol
Boiling Pt:111 °C (1013 hPa)
Melting Pt:–77.8 °C
Density:1.042 g/cm³ (20 °C)
Flash Pt:50 °C
EINECS:204-471-5
UN:2329
ADR:3,III
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
CAS Number:121-45-9
Formula:C₃H₉O₃P
MDL Number:MFCD00008350