Order Entry
Czech Republic
ContactUsLinkComponent
Tributyl phosphate for synthesis, Sigma-Aldrich®
Health
Irritant
Tributyl phosphate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Health
Irritant
Tributyl phosphate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (GC, area%):
    ≥ 99.0 % (a/a)
  • Density (d 20 °C/ 4 °C):
    0.976 - 0.978
  • Identity (IR):
    passes test

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together