Order Entry
Czech Republic
ContactUsLinkComponent
Bismuth(III) nitrate pentahydrate ≥97%, GPR RECTAPUR®
Oxidizing
Irritant
Bismuth(III) nitrate pentahydrate ≥97%, GPR RECTAPUR®
Catalog # 21932.263
Supplier:  VWR Chemicals
CAS Number:  
undefined
Oxidizing
Irritant
Bismuth(III) nitrate pentahydrate ≥97%, GPR RECTAPUR®
Catalog # 21932.263
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    Min. 97 %
  • Alkali and alkaline-earths (as SO4):
    Max. 0.2 %
  • Cl (Chloride):
    Max. 0.02 %
  • SO4 (Sulphate):
    Max. 0.1 %

Specification Test Results

About this item

 :
Boiling Pt:75…80 °C (1013 hPa)
Melting Pt:30 °C
Density:2.736 g/cm³ (20 °C)
ADR:5.1,II
 :485.07 g/mol
EINECS:233-791-8
UN:1477
 :GHS03
 :GHS07
CAS Number:10035-06-0
Formula:Bi(NO₃)₃·5H₂O
MDL Number:MFCD00149157