Diphenyltin dichloride ≥95%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 95% CAS No.: 1135-99-5 Molecular Formula: C12H10Cl2Sn Formula Weight: 343.81 Flash Point: 150°(302°F) Physical Form: Crystalline Sensitivity: Moisture Sensitive Appearance: White Melting Point: 41-43° Boiling Point: 333-337° dec. MDL No.: MFCD00000516
Synonyms: Dichlorodiphenylstannane
- CAS Number:1135-99-5
- Formula:C₁₂H₁₀Cl₂Sn
- MDL Number:MFCD00000516
- Molecular Weight:343.83 g/mol
- Boiling Pt:333…337 °C (dec.)
- Melting Pt:41…43 °C
- Flash Pt:>150 °C (302 °F)
- Storage Temperature:Ambient
- EINECS:214-496-3
- UN:3146
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Purity: 95% CAS No.: 1135-99-5 Molecular Formula: C12H10Cl2Sn Formula Weight: 343.81 Flash Point: 150°(302°F) Physical Form: Crystalline Sensitivity: Moisture Sensitive Appearance: White Melting Point: 41-43° Boiling Point: 333-337° dec. MDL No.: MFCD00000516
Synonyms: Dichlorodiphenylstannane
- CAS Number:1135-99-5
- Formula:C₁₂H₁₀Cl₂Sn
- MDL Number:MFCD00000516
- Molecular Weight:343.83 g/mol
- Boiling Pt:333…337 °C (dec.)
- Melting Pt:41…43 °C
- Flash Pt:>150 °C (302 °F)
- Storage Temperature:Ambient
- EINECS:214-496-3
- UN:3146
- ADR:6.1,III



