
1,2-Diphenyl-1,1,2,2-tetramethyldisilane 97%
Catalog # ACRO387880010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to light brown Low melting mass
- Infrared spectrum:Conforms
- Melting point:33 to 38 °C
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:270.52 g/mol
Boiling Pt:73 °C (0.3 mmHg)
Melting Pt:33…38 °C
Density:0,97 g/cm³ (20 °C)
Flash Pt:105 °C
:GHS07
CAS Number:1145-98-8
Formula:C₁₆H₂₂Si₂
MDL Number:MFCD00054786