

Triethylsilane for synthesis, Sigma-Aldrich®
: Merck
:617-86-7
:
:C₆H₁₆Si
:MFCD00009018
:116.28 g/mol
:107 °C (1013 hPa)
:–156.9 °C
:0.73 g/cm³ (20 °C)
:–3 °C
:1993
:3,II
:M-Clarity™ quality level = MQ200
:210-535-3
:GHS02
:GHS07
:Ambient
- Solubility:(20 °C) (decomposition)
- Molar Mass:116.28 g/mol
- Boiling Point:107 - 109 °C (1013 hPa)
- Vapor Pressure:1 hPa (20 °C)
- Flash Point:-6 °C
- Refractive Index:1.4117 (20 °C, 589 nm)
- Density:0.73 g/cm3 (20 °C)
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Frequently Bought Together