Order Entry
Czech Republic
ContactUsLinkComponent
Tripropyltin chloride for synthesis, Sigma-Aldrich®
Toxic
Environment
Tripropyltin chloride for synthesis, Sigma-Aldrich®
Katalogové číslo 8.08735.0005
Supplier:  Merck
CAS Number:  
Toxic
Environment
Tripropyltin chloride for synthesis, Sigma-Aldrich®
Katalogové číslo 8.08735.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Melting Point:
    -23 °C
  • Molar Mass:
    283.43 g/mol
  • Boiling Point:
    123 °C (17.3 hPa)
  • Flash Point:
    95 °C
  • Density:
    1.29 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :283.43 g/mol
Boiling Pt:234 °C (1013 hPa)
Melting Pt:–23 °C
Density:1.25 g/cm³ (20 °C)
Flash Pt:95 °C
EINECS:218-910-3
UN:2788
ADR:6.1,II
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS09
CAS Number:2279-76-7
Formula:C₉H₂₁ClSn
MDL Number:MFCD00013610