Order Entry
Czech Republic
ContactUsLinkComponent
(3-Aminopropyl)triethoxysilane for synthesis, Sigma-Aldrich®
Corrosive
Irritant
(3-Aminopropyl)triethoxysilane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Irritant
(3-Aminopropyl)triethoxysilane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Molar Mass:
    221.37 g/mol
  • Synonyms:
    3-Aminopropyltriethoxysilane
  • Chemical Formula:
    (C2H5O)3Si(CH2)3NH2

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together