Order Entry
Czech Republic
ContactUsLinkComponent
Vinyltriethoxysilane for synthesis, Sigma-Aldrich®
Vinyltriethoxysilane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Vinyltriethoxysilane for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Molar Mass:
    190.31 g/mol
  • Boiling Point:
    156 °C (1013 hPa)
  • Vapor Pressure:
    7 hPa (20 °C)
  • Flash Point:
    38 °C
  • Refractive Index:
    1.3966 (25 °C, 589 nm)
  • Explosion Limit:
    0.53 - 15 %(V)
  • Density:
    0.91 g/cm3 (20 °C)
  • Ignition Point:
    265 °C

Specification Test Results

Related Information

Frequently Bought Together