

Tin(II) chloride ∼99%, anhydrous, white crystalline powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7772-99-8
:
Formula:SnCl₂
MDL Number:MFCD00011241
:189.62 g/mol
Merck Index:13,08861
Boiling Pt:652 °C (lit.)
Melting Pt:246 °C
Density:3.95 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-868-0
UN:3260
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together